Learn More
Med Vet International Dynarex Xeroburn Burn Dressing, Sterile, 4" X 4", 60/Case
Catalog No. 502830811
Encompass_Preferred
Dynarex XeroBurn™ Burn Dressing is designed to help relieve pain caused by minor burns. This premium dressing is formulated with tea tree oil to provide a cooling effect and help reduce the risk of infection. Designed to temporarily relieve pain from minor burns and scalds Formulated with tea tree oil to help reduce the risk of infection Moisturizing gel helps to cool skin and minimize skin damage   Non-adhesive dressing cushions skin for optimal comfort Ideal for first aid and other medical and homecare settings Not made with natural rubber latex
Catalog No. 50-283-0811 Supplier Med Vet International Supplier No. 1295
Dynarex XeroBurn™ Burn Dressing is designed to help relieve pain caused by minor burns. This premium dressing is formulated with tea tree oil to provide a cooling effect and help reduce the risk of infection. Designed to temporarily relieve pain from minor burns and scalds Formulated with tea tree oil to help reduce the risk of infection Moisturizing gel helps to cool skin and minimize skin damage   Non-adhesive dressing cushions skin for optimal comfort Ideal for first aid and other medical and homecare settings Not made with natural rubber latex