Learn More
Med Vet International Dynarex Sterile Dynaguard Waterproof Composite Dressing, 4" X 4", 120/Case
Supplier:  Med Vet International 3035
Encompass_Preferred
Dynarex’s DynaGuard Waterproof Dressing is a sterile, multi-layered foam dressing that provides a warm moist healing environment. The waterproof top layer acts as a bacterial barrier, protecting the wound from any outside liquids that can cause infection. The inner foam layer absorbs drainage, while the low-adherent contact layer and adhesive non-woven fabric border provide protection to the wound bed and are gentle on the skin. Multi-layer, medium-absorbency dressing used for moderate-exuding wounds Helps provide a moist healing environment with protection from outside contaminants Medium-absorbency dressing used for moderate-exuding wounds Four-layer construction: low-adherent contact layer, absorbent pad, semi-occlusive barrier film and conformable, adhesive non-woven fabric border Primary or secondary dressing to manage chronic and acute wounds at stages 2, 3, and 4
Catalog No. 50-283-1193
 Restricted Item