Learn More
Med Vet International Dynarex Sterile Oil Emulsion Dressing, 3" X 8", 144/Case
Catalog No. 502830849
Encompass_Preferred
A non-adherent knitted gauze impregnated with a petrolatum and sunflower oil in an emulsion blend. These conformable, non-occlusive dressings allow exudate to flow away from the wound into a secondary dressing, minimizing skin maceration. Knitted gauze dressings, evenly impregnated with an emulsion blend of petrolatum and sunflower oil. Allow exudate to flow away from the wound, minimizing the risk of skin maceration. Non-occlusive, primary dressing used for low exuding wounds at stages 2, 3 or 4.
Catalog No. 50-283-0849 Supplier Med Vet International Supplier No. 3068
A non-adherent knitted gauze impregnated with a petrolatum and sunflower oil in an emulsion blend. These conformable, non-occlusive dressings allow exudate to flow away from the wound into a secondary dressing, minimizing skin maceration. Knitted gauze dressings, evenly impregnated with an emulsion blend of petrolatum and sunflower oil. Allow exudate to flow away from the wound, minimizing the risk of skin maceration. Non-occlusive, primary dressing used for low exuding wounds at stages 2, 3 or 4.