Learn More
Med Vet International Dynarex Sterile Siligentle Silicone Bordered Foam Dressing, 4" X 4", 120/Case
Catalog No. 502829450
Encompass_Preferred
Highly absorbent dressings with a vapor permeable waterproof film to lock away exudate to minimize pain and trauma. Provides a warm, moist healing environment to help prevent bacterial contamination. Soft and comfortable with superior absorption locks exudate away. Vapor permeable waterproof film and silicone contact layer that is gentle on wounds and moves without losing adherence. High absorbency dressing used for high exuding wounds. Primary or secondary dressing to treat and manage partial to full thickness wounds at stages 2, 3 or 4.
Catalog No. 50-282-9450 Supplier Med Vet International Supplier No. DYN3059
Highly absorbent dressings with a vapor permeable waterproof film to lock away exudate to minimize pain and trauma. Provides a warm, moist healing environment to help prevent bacterial contamination. Soft and comfortable with superior absorption locks exudate away. Vapor permeable waterproof film and silicone contact layer that is gentle on wounds and moves without losing adherence. High absorbency dressing used for high exuding wounds. Primary or secondary dressing to treat and manage partial to full thickness wounds at stages 2, 3 or 4.