Learn More
Med Vet International Dynarex Sterile Dynafoam Waterproof Bordered Foam Dressing, 6" X 6", 120/Case
Catalog No. 502829671
Encompass_Preferred
A sterile, waterproof adhesive foam that provides a warm moist healing environment. The multi-layer design helps protect and prevent bacterial contamination while promoting healing. Maintains a warm, moist healing environment to help prevent bacterial contamination, reduce pain and promote faster healing. Multi-layer dressing with low adherent contact layer that helps protect the wound bed and waterproof top layer which protects from outside fluids. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to manage chronic and acute wounds at stages 2, 3 and 4. Used on lower extremity ulcers, donor sites and partial to full thickness wounds.
Catalog No. 50-282-9671 Supplier Med Vet International Supplier No. 3038
A sterile, waterproof adhesive foam that provides a warm moist healing environment. The multi-layer design helps protect and prevent bacterial contamination while promoting healing. Maintains a warm, moist healing environment to help prevent bacterial contamination, reduce pain and promote faster healing. Multi-layer dressing with low adherent contact layer that helps protect the wound bed and waterproof top layer which protects from outside fluids. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to manage chronic and acute wounds at stages 2, 3 and 4. Used on lower extremity ulcers, donor sites and partial to full thickness wounds.