Learn More
Med Vet International Dynarex Sterile Curafoam Foam Dressing, 4" X 4.25", 120/Case
Catalog No. 502829408
Encompass_Preferred
A non-bordered, sterile dressing used to manage and treat chronic and acute partial and full thickness wounds. The dressing provides a warm, moist and cushioned healing environment. Maintains a warm, moist and cushioned healing environment. Prevents bacterial contamination, reducing pain and promoting faster healing. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to manage chronic and acute wounds at stages 2, 3 and 4. Used on lower extremity ulcers, donor sites and partial to full thickness wounds.
Catalog No. 50-282-9408 Supplier Med Vet International Supplier No. DYN3012
A non-bordered, sterile dressing used to manage and treat chronic and acute partial and full thickness wounds. The dressing provides a warm, moist and cushioned healing environment. Maintains a warm, moist and cushioned healing environment. Prevents bacterial contamination, reducing pain and promoting faster healing. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to manage chronic and acute wounds at stages 2, 3 and 4. Used on lower extremity ulcers, donor sites and partial to full thickness wounds.