Learn More
Med Vet International Dynarex Sterile Siligentle Non-Adhesive Silicone Foam Dressing, 6" X 6", 120/Case
Catalog No. 502831944
Encompass_Preferred
Highly absorbent dressings with a vapor permeable waterproof film to lock away exudate to minimize pain and trauma. Provides a warm, moist healing environment to help prevent bacterial contamination. Soft and comfortable with superior absorption locks exudate away. Vapor permeable waterproof film and silicone contact layer that is gentle on wounds and moves without losing adherence. High absorbency dressing used for high exuding wounds. Primary or secondary dressing to treat and manage partial to full thickness wounds at stages 2, 3 or 4.
Catalog No. 50-283-1944 Supplier Med Vet International Supplier No. DYN3057
Highly absorbent dressings with a vapor permeable waterproof film to lock away exudate to minimize pain and trauma. Provides a warm, moist healing environment to help prevent bacterial contamination. Soft and comfortable with superior absorption locks exudate away. Vapor permeable waterproof film and silicone contact layer that is gentle on wounds and moves without losing adherence. High absorbency dressing used for high exuding wounds. Primary or secondary dressing to treat and manage partial to full thickness wounds at stages 2, 3 or 4.