Learn More
Med Vet International Foam Dressing ComfortFoam™ Border 9 X 9 Inch With Border Waterproof Backing Silicone Adhesive Sacral Sterile, 43990, 5/Box
Catalog No. 503045365
Encompass_Preferred
The 9- x 9-inch DermaRite ComfortFoam border facilitates faster and proper healing of wounds A multilayer design optimizes wound protection, thermal insulation and absorbency The skin-friendly adhesive foam dressing sticks well and removes easily A low-profile border improves adhesion and flexibility for application in difficult areas
Catalog No. 50-304-5365 Supplier Med Vet International Supplier No. 946565BX
The 9- x 9-inch DermaRite ComfortFoam border facilitates faster and proper healing of wounds A multilayer design optimizes wound protection, thermal insulation and absorbency The skin-friendly adhesive foam dressing sticks well and removes easily A low-profile border improves adhesion and flexibility for application in difficult areas