Learn More
Med Vet International Dynarex Sterile Foamflex Foam Dressing, Non-Adhesive, Waterproof, 6" X 6", 120/Case
Catalog No. 502831279
Encompass_Preferred
Non-Adhesive Waterproof foam dressings provide a comfortable, warm, moist wound environment and is made with a semi-permeable film to help prevent external bacterial contamination. The foam is flexible and conforms to hard to dress areas. Maintains a warm, moist wound environment to help prevent bacterial contamination. Semi-permeable film bonds to the foam, allowing oxygen to pass through, letting the wound breathe. Flexible and easy to conform to hard-to-dress areas. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to manage chronic and acute wounds at stages 2, 3 and 4. Used on lower extremity ulcers, donor sites and partial to full thickness wounds.
Catalog No. 50-283-1279 Supplier Med Vet International Supplier No. 3025
Non-Adhesive Waterproof foam dressings provide a comfortable, warm, moist wound environment and is made with a semi-permeable film to help prevent external bacterial contamination. The foam is flexible and conforms to hard to dress areas. Maintains a warm, moist wound environment to help prevent bacterial contamination. Semi-permeable film bonds to the foam, allowing oxygen to pass through, letting the wound breathe. Flexible and easy to conform to hard-to-dress areas. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to manage chronic and acute wounds at stages 2, 3 and 4. Used on lower extremity ulcers, donor sites and partial to full thickness wounds.