Learn More
Med Vet International Dynarex Petroleum Gauze Dressing, 3" X 9", 200/Case
Catalog No. 502831116
Encompass_Preferred
Dynarex’s Petroleum Gauze is a lightweight, gauze dressing impregnated with petroleum jelly, helping maintain a moist wound environment for autolytic healing. The dressing conforms closely to the wound bed and will not adhere upon removal. Petroleum Gauze is used as a primary dressing on low draining wounds of full thickness such as, donor sites, minor burns, tube sites and non-infected wounds at stages 2, 3 and 4. A non-adherent knitted gauze, impregnated with petroleum jelly to help maintain a moist wound environment for autolytic healing.
Catalog No. 50-283-1116 Supplier Med Vet International Supplier No. 3039
Dynarex’s Petroleum Gauze is a lightweight, gauze dressing impregnated with petroleum jelly, helping maintain a moist wound environment for autolytic healing. The dressing conforms closely to the wound bed and will not adhere upon removal. Petroleum Gauze is used as a primary dressing on low draining wounds of full thickness such as, donor sites, minor burns, tube sites and non-infected wounds at stages 2, 3 and 4. A non-adherent knitted gauze, impregnated with petroleum jelly to help maintain a moist wound environment for autolytic healing.