Learn More
Med Vet International Dynarex Sterile Xeroform Gauze Dressing, 5" X 9", 100/Case
Catalog No. 502831280
Encompass_Preferred
A non-adherent knitted gauze impregnated with a blend of petroleum, 3% Bismuth, mineral, and paraffin oils. These highly conformable, non-occlusive dressings allow the wounds to breathe and are indicated for use in deep to shallow wounds with minimal exudate. Helps maintain a moist wound environment Non-adherent and non-occlusive Conformable for use in deep and shallow wounds Knitted gauze, impregnated wound dressing Designed to allow the wound to breathe, minimizing skin and tissue maceration Primary dressing on low-exuding wounds at stages 2, 3, or 4 Active ingredient 3% Bismuth reduces wound odor
Catalog No. 50-283-1280 Supplier Med Vet International Supplier No. 3054
A non-adherent knitted gauze impregnated with a blend of petroleum, 3% Bismuth, mineral, and paraffin oils. These highly conformable, non-occlusive dressings allow the wounds to breathe and are indicated for use in deep to shallow wounds with minimal exudate. Helps maintain a moist wound environment Non-adherent and non-occlusive Conformable for use in deep and shallow wounds Knitted gauze, impregnated wound dressing Designed to allow the wound to breathe, minimizing skin and tissue maceration Primary dressing on low-exuding wounds at stages 2, 3, or 4 Active ingredient 3% Bismuth reduces wound odor