Learn More
Med Vet International Dynarex Hemostat Gauze Dressing, 2" X 4", 200/Case
Catalog No. 502831328
Encompass_Preferred
The Dynarex DynaSafety Hemostat Gauze Dressing is a premium water-soluble dressing designed to help protect wounds and manage bleeding. The Dressing contains a topical compound that coverts to a glucose gel upon contact with blood, providing direct pressure on blood vessels to help reduce bleeding. The Dynarex Hemostat Gauze Dressing is ideal for wounds, small cuts and abrasions, lacerations, and punctures.  The Dynarex DynaSafety Hemostat Gauze Dressing is a high-quality bandage designed to help manage bleeding. The water-soluble Dressing forms a glucose gel when in contact with blood to help protect the wound area. 
Catalog No. 50-283-1328 Supplier Med Vet International Supplier No. 3686
The Dynarex DynaSafety Hemostat Gauze Dressing is a premium water-soluble dressing designed to help protect wounds and manage bleeding. The Dressing contains a topical compound that coverts to a glucose gel upon contact with blood, providing direct pressure on blood vessels to help reduce bleeding. The Dynarex Hemostat Gauze Dressing is ideal for wounds, small cuts and abrasions, lacerations, and punctures.  The Dynarex DynaSafety Hemostat Gauze Dressing is a high-quality bandage designed to help manage bleeding. The water-soluble Dressing forms a glucose gel when in contact with blood to help protect the wound area. 