Learn More
Med Vet International Dynarex Dynaderm - Hydrocolloid Dressing - Extra Thin, 4" X 4", 120/Case
Supplier:  Med Vet International 3018
Encompass_Preferred
DynaDerm™ Hydrocolloid Dressings are flexible, insulated coverings designed to help shield non-infected, partial, and full-thickness wounds including pressure ulcers, abrasions, and donor sites. These semi-occlusive dressings help to accelerate the healing process by preserving moisture and promoting autolytic debridement. The Hydrocolloid Dressings’ mild adhesive surface gently adheres to surrounding skin but won’t stick to the wound bed for optimal comfort and bandage longevity. These convenient Dressings can be safely worn for several days depending on wound condition and exudate levels.  DynaDerm™ Hydrocolloid Dressings are high-quality sterile dressings designed to safely and efficiently protect a wide variety of non-infected wounds from external contamination. These premium Dressings help to support an optimal healing environment by providing thermal insulation while maintaining natural moisture. 
Catalog No. 50-282-9689