Learn More
Med Vet International Dynarex Sterile Dynaderm Hydrocolloid Dressing, Extra Thin, 6" X 6", 60/Case
Catalog No. 502831300
Encompass_Preferred
Hydrocolloid Dressings provide a sterile, moist, insulated healing environment. They are used on uninfected, partial or full thickness wounds with low amounts of exudate. Provides a moist insulated healing environment permeable to moisture, vapor and oxygen. Promotes faster healing for uninfected wounds. Lower absorbency level, best suited for pressure ulcers/injuries at stages 2, 3 and 4. Primary or secondary dressing to manage partial and full thickness wounds.
Catalog No. 50-283-1300 Supplier Med Vet International Supplier No. 3019
Add to Cart
Add to Cart
Hydrocolloid Dressings provide a sterile, moist, insulated healing environment. They are used on uninfected, partial or full thickness wounds with low amounts of exudate. Provides a moist insulated healing environment permeable to moisture, vapor and oxygen. Promotes faster healing for uninfected wounds. Lower absorbency level, best suited for pressure ulcers/injuries at stages 2, 3 and 4. Primary or secondary dressing to manage partial and full thickness wounds.