Learn More
Med Vet International Dynarex Island Dressing, Sterile, (Individually Bagged), 2" X 3.5", 300/Case
Catalog No. 502830847
Encompass_Preferred
Island dressings are non-adherent wound contact layers, individually packed for convenience. Ideal for use as a cover dressing after packing large, deep wounds Individually packed in peel open pouches Provides excellent absorption and cushioning protection Soft, non-stick dressing with adhesive border Not made with natural rubber latex
Catalog No. 50-283-0847 Supplier Med Vet International Supplier No. 3492
Island dressings are non-adherent wound contact layers, individually packed for convenience. Ideal for use as a cover dressing after packing large, deep wounds Individually packed in peel open pouches Provides excellent absorption and cushioning protection Soft, non-stick dressing with adhesive border Not made with natural rubber latex