Learn More
Med Vet International IV Dressing 3M Tegaderm Film 1-1/2 X 1-3/4 inch Sterile, 1680, 400/Case
Catalog No. 502832022
Encompass_Preferred
Designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture A cost-effective I.V. and central line catheter securement solution Innovative adhesives balance securement and gentle removal Safe to wear up to 7 days for CVC's and may be worn for the life of the PIV catheter A waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses Transparent / White
Catalog No. 50-283-2022 Supplier Med Vet International Supplier No. 1051704CS
Designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture A cost-effective I.V. and central line catheter securement solution Innovative adhesives balance securement and gentle removal Safe to wear up to 7 days for CVC's and may be worn for the life of the PIV catheter A waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses Transparent / White