Learn More
Med Vet International Foam Dressing Kerrafoam Gentle Border 4 X 8 inch With Border Film Backing Silicone Adhesive Rectangle Sterile, Cwl1012, 50/Case
Catalog No. 502828953
Encompass_Preferred
The smooth outer film layer helps to reduce shear and friction, create a water- resistant bacterial barrier, and maintain an ideal MVTR A soft inner pouch allows the layers to move freely, absorbing shear and friction forces that can damage peri-wound skin Absorbent core and foam pad combine to lock in exudate , bacteria and MMPs - even under compression A soft silicone layer ensures gentle adhesion, excellent absorption rate and non traumatic removal Horizontal and vertical wicking layer efficiently increases absorption capacity distributing exudate throughout the dressing
Catalog No. 50-282-8953 Supplier Med Vet International Supplier No. 1189333CS
The smooth outer film layer helps to reduce shear and friction, create a water- resistant bacterial barrier, and maintain an ideal MVTR A soft inner pouch allows the layers to move freely, absorbing shear and friction forces that can damage peri-wound skin Absorbent core and foam pad combine to lock in exudate , bacteria and MMPs - even under compression A soft silicone layer ensures gentle adhesion, excellent absorption rate and non traumatic removal Horizontal and vertical wicking layer efficiently increases absorption capacity distributing exudate throughout the dressing