Learn More
Med Vet International Foam Dressing Kerrafoam Gentle Border 7 X 8 inch With Border Film Backing Silicone Adhesive Oval Sterile, Cwl1135, 90/Case
Catalog No. 502828954
Encompass_Preferred
Smooth outer film layer helps to reduce shear and friction, create a water-resistant bacterial barrier, and maintain an ideal MVTR Soft inner pouch allows the layers to glide, absorbing shear and friction forces that can damage peri-wound skin Absorbent core and foam pad combine to lock in exudate, bacteria and MMPs – even under compression Soft silicone layer ensures gentle adhesion, excellent absorption rate and nontraumatic removal Horizontal and vertical wicking layer efficiently increases absorption capacity distributing exudate throughout the dressing Silicone foam dressing with film backing is oval and is 18 x 21 centimeters
Catalog No. 50-282-8954 Supplier Med Vet International Supplier No. 1189338CS
Smooth outer film layer helps to reduce shear and friction, create a water-resistant bacterial barrier, and maintain an ideal MVTR Soft inner pouch allows the layers to glide, absorbing shear and friction forces that can damage peri-wound skin Absorbent core and foam pad combine to lock in exudate, bacteria and MMPs – even under compression Soft silicone layer ensures gentle adhesion, excellent absorption rate and nontraumatic removal Horizontal and vertical wicking layer efficiently increases absorption capacity distributing exudate throughout the dressing Silicone foam dressing with film backing is oval and is 18 x 21 centimeters