Learn More
Med Vet International Hydrogel Wound Dressing Kerralite Cool Border Sheet 4 X 4 inch Sheet Sterile, Cwl1008, 260/Case
Catalog No. 502830735
Encompass_Preferred
Soothing, debriding and moisture-balancing dressings Adhesive border enables the dressing to be secured in position Suitable for many wound types Helps to improve healing of low exuding wounds Increases granulation tissue of chronic wounds Limits the growth of bacteria Soothing and cooling effect minimize discomfort, especially at dressing changes KerraLite Cool won't dry out or stick to the underlying tissue on removal, reducing the risk of maceration Protects against wound dehydration 4 x 4 inches
Catalog No. 50-283-0735 Supplier Med Vet International Supplier No. 1189331CS
Soothing, debriding and moisture-balancing dressings Adhesive border enables the dressing to be secured in position Suitable for many wound types Helps to improve healing of low exuding wounds Increases granulation tissue of chronic wounds Limits the growth of bacteria Soothing and cooling effect minimize discomfort, especially at dressing changes KerraLite Cool won't dry out or stick to the underlying tissue on removal, reducing the risk of maceration Protects against wound dehydration 4 x 4 inches