Learn More
Med Vet International Hydrogel Wound Dressing Kerralite Cool Border Sheet 6 X 6 inch Sheet Sterile, Cwl1009, 160/Case
Catalog No. 502829230
Encompass_Preferred
Hydrogel moisture balancing dressing with Pro-inoic™ technology 3M™ Kerralite Cool™ Border Dressings comprise a polyurethane outer film, a Pro-ionic™5 gel sheet and a gentle adhesive border The dressings are designed to actively respond to wound conditions by donating or absorbing fluid Suitable for use under compression Waterproof 7 day wear time
Catalog No. 50-282-9230 Supplier Med Vet International Supplier No. 1189365CS
Hydrogel moisture balancing dressing with Pro-inoic™ technology 3M™ Kerralite Cool™ Border Dressings comprise a polyurethane outer film, a Pro-ionic™5 gel sheet and a gentle adhesive border The dressings are designed to actively respond to wound conditions by donating or absorbing fluid Suitable for use under compression Waterproof 7 day wear time