Learn More
Med Vet International Adhesive Dressing 3M Medipore 2 X 2-3/4 inch Soft Cloth Rectangle White Sterile, 3562, 50/Box
Catalog No. 502829600
Encompass_Preferred
3M's Medipore +Pad soft cloth adhesive wound dressings support the healing of wounds, abrasions and surgical incisions Soft, porous material absorbs drainage and is breathable for improved airflow and comfort Dressings adhere to skin without sticking to wounds, making removal easy and safe Gentle flexibility conforms to bodily contours, even as body areas move or if swelling occurs Individually wrapped, the Medipore wound dressings remain sterile before use Sterile adhesive dressings are 2 x 2-3/4 inches with an absorbent pad of 1 x 1-1/2 inches
Catalog No. 50-282-9600 Supplier Med Vet International Supplier No. 324093BX
3M's Medipore +Pad soft cloth adhesive wound dressings support the healing of wounds, abrasions and surgical incisions Soft, porous material absorbs drainage and is breathable for improved airflow and comfort Dressings adhere to skin without sticking to wounds, making removal easy and safe Gentle flexibility conforms to bodily contours, even as body areas move or if swelling occurs Individually wrapped, the Medipore wound dressings remain sterile before use Sterile adhesive dressings are 2 x 2-3/4 inches with an absorbent pad of 1 x 1-1/2 inches