Learn More
Med Vet International Adhesive Dressing 3M Medipore 3-1/2 X 8 inch Soft Cloth Rectangle White Sterile, 3570, 100/Case
Catalog No. 502829890
Encompass_Preferred
Soft, porous and breathable. nonadherent pad won't stick to the wound, thus minimizing disruption of the healing tissues upon removal Absorbent pad protects wound and absorbs drainage, while allowing skin to breathe, thus reducing the potential for maceration around the wound edges Helps reduce the risk of skin maceration Dressing provides excellent adhesion, yet is gentle to the skin Conforms to body contours 3-1/2 x 8 inch adhesive dressing has a 1-3/4 x 6 inch pad
Catalog No. 50-282-9890 Supplier Med Vet International Supplier No. 324095CS
Soft, porous and breathable. nonadherent pad won't stick to the wound, thus minimizing disruption of the healing tissues upon removal Absorbent pad protects wound and absorbs drainage, while allowing skin to breathe, thus reducing the potential for maceration around the wound edges Helps reduce the risk of skin maceration Dressing provides excellent adhesion, yet is gentle to the skin Conforms to body contours 3-1/2 x 8 inch adhesive dressing has a 1-3/4 x 6 inch pad