Learn More
Med Vet International Adhesive Dressing 3M Medipore 3-1/2 X 13-3/4 inch Soft Cloth Rectangle White Sterile, 3573, 25/Box
Catalog No. 502829829
Encompass_Preferred
Enhances comfort: Soft, porous and breathable. nonadherent pad won't stick to the wound, thus minimizing disruption of the healing tissues upon removal Helps support healing: Absorbent pad protects wound and absorbs drainage, while allowing skin to breathe, thus reducing the potential for maceration around the wound edges Helps reduce the risk of skin maceration Gentle to the skin: Dressing provides excellent adhesion, yet is gentle to the skin Conforms to body contours 3-1/2 x 13-3/4 inch adhesive dressing has a 1-3/4 x 11-3/4 inch pad
Catalog No. 50-282-9829 Supplier Med Vet International Supplier No. 324097BX
Enhances comfort: Soft, porous and breathable. nonadherent pad won't stick to the wound, thus minimizing disruption of the healing tissues upon removal Helps support healing: Absorbent pad protects wound and absorbs drainage, while allowing skin to breathe, thus reducing the potential for maceration around the wound edges Helps reduce the risk of skin maceration Gentle to the skin: Dressing provides excellent adhesion, yet is gentle to the skin Conforms to body contours 3-1/2 x 13-3/4 inch adhesive dressing has a 1-3/4 x 11-3/4 inch pad