Learn More
Med Vet International Adhesive Dressing 3M Medipore 3-1/2 X 6 inch Soft Cloth Rectangle White Sterile, 3569, 25/Box
Catalog No. 502830876
Encompass_Preferred
3M's Medipore pad covers and protects different wound types for versatile use The white pad is highly absorbent to handle wound drainage effectively It won't stick to the wound bed for less pain upon removal A soft cloth tape backing provides superior breathability The dressing is conformable for easy handling and application 3-1/2 x 6 inch adhesive dressing has a 1-3/4 x 4 inch pad
Catalog No. 50-283-0876 Supplier Med Vet International Supplier No. 324094BX
3M's Medipore pad covers and protects different wound types for versatile use The white pad is highly absorbent to handle wound drainage effectively It won't stick to the wound bed for less pain upon removal A soft cloth tape backing provides superior breathability The dressing is conformable for easy handling and application 3-1/2 x 6 inch adhesive dressing has a 1-3/4 x 4 inch pad