Learn More
Med Vet International Adhesive Dressing 3M Medipore 6 X 6 inch Soft Cloth Square White Sterile, 3568, Each
Catalog No. 502830858
Encompass_Preferred
The 3M Medipore pad is porous, allowing the skin to breathe It is gentle to the skin to enhance comfort The pad flexes with body movement and swelling so that it can easily conform to contours The absorbent pad is nonadherent and won't disrupt the wound bed upon removal 6 x 6 inch dressing with 4 x 4-1/2 inch pad
Catalog No. 50-283-0858 Supplier Med Vet International Supplier No. 378957EA
The 3M Medipore pad is porous, allowing the skin to breathe It is gentle to the skin to enhance comfort The pad flexes with body movement and swelling so that it can easily conform to contours The absorbent pad is nonadherent and won't disrupt the wound bed upon removal 6 x 6 inch dressing with 4 x 4-1/2 inch pad