Learn More
Med Vet International Transparent Film Dressing With Pad Nexcare Tegaderm+ Pad 2-3/8 X 4 inch 1 Tab DelIVery Rectangle Sterile, 05113183558, 5/Box
Catalog No. 502831454
Encompass_Preferred
A waterproof dressing which consists of a non-stick, absorbent pad bonded to a larger thin film backing coated with a hypoallergenic, water resistant adhesive Use to protect wounds such as cuts, burns, abrasions and post-surgical incisions Non-stick pad with clear adhesive cover
Catalog No. 50-283-1454 Supplier Med Vet International Supplier No. 863879BX
A waterproof dressing which consists of a non-stick, absorbent pad bonded to a larger thin film backing coated with a hypoallergenic, water resistant adhesive Use to protect wounds such as cuts, burns, abrasions and post-surgical incisions Non-stick pad with clear adhesive cover