Learn More
Med Vet International Dynarex View Guard Transparent Film Dressing Roll, 2" X 11 Yd, 12/Case
Catalog No. 502832139
Encompass_Preferred
Dynarex View Guard Transparent Film Dressing Rolls are premium transparent dressings designed to cover and protect skin after tattoo needlework. Crafted from flexible materials, these Dressing Rolls conform easily to body contours to allow movement without stress or irritation to skin. These versatile Rolls can be easily cut to the desired size or shape to suit a variety of needs. Dynarex View Guard Transparent Film Dressing Rolls are ideal for use around tattoos, surgical sites, and incisions.  Dynarex View Guard Transparent Film Dressing Rolls are designed from flexible materials to conform to body contours, allowing movement without stressing skin. The Rolls can be easily cut to desired shape or size for a variety of applications and needs. 
Catalog No. 50-283-2139 Supplier Med Vet International Supplier No. 3639
Dynarex View Guard Transparent Film Dressing Rolls are premium transparent dressings designed to cover and protect skin after tattoo needlework. Crafted from flexible materials, these Dressing Rolls conform easily to body contours to allow movement without stress or irritation to skin. These versatile Rolls can be easily cut to the desired size or shape to suit a variety of needs. Dynarex View Guard Transparent Film Dressing Rolls are ideal for use around tattoos, surgical sites, and incisions.  Dynarex View Guard Transparent Film Dressing Rolls are designed from flexible materials to conform to body contours, allowing movement without stressing skin. The Rolls can be easily cut to desired shape or size for a variety of applications and needs. 