Learn More
Med Vet International Dynarex Sterile Dynaginate Calcium Alginate Rope Dressing, 2 G X 30 Cm, 60/Case
Catalog No. 502831124
Encompass_Preferred
Calcium Alginate Dressings contours to wound bed and changes to a fibrous gel when in contact with wound exudate, creating an optimal moist wound environment important to healing. Changes to a fibrous gel when in contact with wound exudate, creating a moist wound environment. Contours to the wound bed and will not adhere to the wound upon removal. High absorbency dressing used for heavily exuding wounds. Primary dressing ideal for the management of partial and full thickness wounds at stages 3 or 4.
Catalog No. 50-283-1124 Supplier Med Vet International Supplier No. 3029
Calcium Alginate Dressings contours to wound bed and changes to a fibrous gel when in contact with wound exudate, creating an optimal moist wound environment important to healing. Changes to a fibrous gel when in contact with wound exudate, creating a moist wound environment. Contours to the wound bed and will not adhere to the wound upon removal. High absorbency dressing used for heavily exuding wounds. Primary dressing ideal for the management of partial and full thickness wounds at stages 3 or 4.