Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 2-3/4 X 2-3/4 inch With Border Film Backing Acrylic Adhesive Square Sterile, 90615, 40/Case
Catalog No. 502830461
Encompass_Preferred
3M's Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply to fingers and toes, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering The white foam dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This 2¾ x 2¾ inch dressing will not stick to wounds or disrupt healing tissue
Catalog No. 50-283-0461 Supplier Med Vet International Supplier No. 695851CS
3M's Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply to fingers and toes, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering The white foam dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This 2¾ x 2¾ inch dressing will not stick to wounds or disrupt healing tissue