Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 2 X 2 inch Without Border Film Backing NonAdhesive Square Sterile, 90600, 40/Case
Catalog No. 502830928
Encompass_Preferred
Multi-layer design provides high absorbency with high breathability to reduce the risk of maceration Adapts to changing levels of exudate to maintain moisture balance for optimal wound healing Particularly suited for situations where increased securement or longer wear time is needed Conformable for difficult body contours
Catalog No. 50-283-0928 Supplier Med Vet International Supplier No. 575882CS
Multi-layer design provides high absorbency with high breathability to reduce the risk of maceration Adapts to changing levels of exudate to maintain moisture balance for optimal wound healing Particularly suited for situations where increased securement or longer wear time is needed Conformable for difficult body contours