Learn More
Med Vet International Transparent Film Dressing With Pad 3M Tegaderm +Pad 3-1/2 X 4 inch Frame Style DelIVery Rectangle Sterile, 3586, 100/Case
Catalog No. 502829510
Encompass_Preferred
Sterile film dressing provides an effective bacterial and viral barrier Conforms to body contours and flexes with movement or swelling Can be worn up to 4 days Waterproof film allows patients to shower without removing the dressing if the dressing is intact Nonadherent pad will not stick to the wound, allowing normal wound healing with less pain and trauma Breathable film allows skin to function normally with good exchange of moisture vapor and oxygen 3½ x 4 inches
Catalog No. 50-282-9510 Supplier Med Vet International Supplier No. 318721CS
Sterile film dressing provides an effective bacterial and viral barrier Conforms to body contours and flexes with movement or swelling Can be worn up to 4 days Waterproof film allows patients to shower without removing the dressing if the dressing is intact Nonadherent pad will not stick to the wound, allowing normal wound healing with less pain and trauma Breathable film allows skin to function normally with good exchange of moisture vapor and oxygen 3½ x 4 inches