Learn More
Med Vet International Foam Dressing 3M Tegaderm 4 X 4-1/4 inch Without Border Film Backing Silicone Face Square Sterile, 90631, 40/Case
Catalog No. 502830201
Encompass_Preferred
Unique multi-layer design absorbs and evaporates moisture while allowing exudate to pass through to help maintain a moist wound environment that supports healing Fluid management properties of the dressing prevent pooling or migration, which helps reduce the potential for skin maceration Strong and gentle adhesive minimizes disruption to skin during removal Secure with elastic or cohesive wrap, tape or other methods as needed Suitable for use on fragile skin and with compression therapy Strong and gentle adhesive minimizes disruption to skin during removal
Catalog No. 50-283-0201 Supplier Med Vet International Supplier No. 1078884CS
Unique multi-layer design absorbs and evaporates moisture while allowing exudate to pass through to help maintain a moist wound environment that supports healing Fluid management properties of the dressing prevent pooling or migration, which helps reduce the potential for skin maceration Strong and gentle adhesive minimizes disruption to skin during removal Secure with elastic or cohesive wrap, tape or other methods as needed Suitable for use on fragile skin and with compression therapy Strong and gentle adhesive minimizes disruption to skin during removal