Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 4 X 4-3/4 inch Frame Style DelIVery Rectangle Sterile, 1626, 200/Case
Catalog No. 502831960
Encompass_Preferred
Tegaderm film allows you to easily view the wound site Its breathable film allows for the exchange of moisture to hasten healing This Tegaderm dressing is flexible and moves with the body for added comfort This bandage is waterproof and keeps out contaminants including viruses and bacteria Transparent film dressing has a frame-style delivery system that makes placement accurate and easy 4 x 4-3/4 inches
Catalog No. 50-283-1960 Supplier Med Vet International Supplier No. 277404CS
Tegaderm film allows you to easily view the wound site Its breathable film allows for the exchange of moisture to hasten healing This Tegaderm dressing is flexible and moves with the body for added comfort This bandage is waterproof and keeps out contaminants including viruses and bacteria Transparent film dressing has a frame-style delivery system that makes placement accurate and easy 4 x 4-3/4 inches