Learn More
Med Vet International IV Specialty Dressing 3M Tegaderm IV Advanced Adhesive / Film 4 X 6-1/8 inch Sterile, 1689, 25/Box
Catalog No. 502832142
Encompass_Preferred
A cost-effective I.V. and central line catheter securement solution, 3M™ Tegaderm™ I.V. advanced securement dressing is designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture Safe to wear up to 7 days for CVCs (central venous catheter) and may be worn for the life of the PIV (peripheral venous) catheter Innovative adhesives balance securement and gentle removal Easy to apply and remove A waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses 4 x 6-1/8 inches
Catalog No. 50-283-2142 Supplier Med Vet International Supplier No. 835844BX
A cost-effective I.V. and central line catheter securement solution, 3M™ Tegaderm™ I.V. advanced securement dressing is designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture Safe to wear up to 7 days for CVCs (central venous catheter) and may be worn for the life of the PIV (peripheral venous) catheter Innovative adhesives balance securement and gentle removal Easy to apply and remove A waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses 4 x 6-1/8 inches