Learn More
Med Vet International Alginate Dressing 3M Tegaderm High integrity 4 X 4 inch Square, 90112, 50/Case
Catalog No. 502831080
Encompass_Preferred
Gel forming for optimal moist wound healing environment Sterile, highly conformable, and absorbent primary wound care dressings for use on moderately to heavily exudating wound High integrity and high gelling alginate dressing that maintains a moist wound healing environment
Catalog No. 50-283-1080 Supplier Med Vet International Supplier No. 318649CS
Gel forming for optimal moist wound healing environment Sterile, highly conformable, and absorbent primary wound care dressings for use on moderately to heavily exudating wound High integrity and high gelling alginate dressing that maintains a moist wound healing environment