Learn More
Med Vet International Foam Dressing 3M Tegaderm 6-1/2 X 6-1/2 inch With Border Film Backing Silicone Adhesive Heel / Contour Sterile, 90646, 20/Case
Catalog No. 502830311
Encompass_Preferred
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 6-1/2 x 6-1/2 inch silicone foam dressing is particularly well suited for heels
Catalog No. 50-283-0311 Supplier Med Vet International Supplier No. 1078887CS
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 6-1/2 x 6-1/2 inch silicone foam dressing is particularly well suited for heels