Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 3-1/2 X 3-1/2 inch Without Border Film Backing NonAdhesive Fenestrated Square Sterile, 90604, 40/Case
Catalog No. 502831165
Encompass_Preferred
Provides a water resistant seal while dressing is intact Allows showering Dressing absorbs/manages low to high levels of exudate, protects peri-wound skin and maintains moisture balance Gentle, absorbent and secure silicone foam border dressing easily handles high to low exudating wounds Stays in place, yet removes gently, making it a good choice for fragile skin Adhesive film border is thin and has a low profile to minimize edge roll Dressing does not adhere to the wound bed and peri-wound skin (undisturbed healing environment) Can be worn for up to 7 days on skin and compromised skin 1½ x 1½ inch pad
Catalog No. 50-283-1165 Supplier Med Vet International Supplier No. 459162CS
Provides a water resistant seal while dressing is intact Allows showering Dressing absorbs/manages low to high levels of exudate, protects peri-wound skin and maintains moisture balance Gentle, absorbent and secure silicone foam border dressing easily handles high to low exudating wounds Stays in place, yet removes gently, making it a good choice for fragile skin Adhesive film border is thin and has a low profile to minimize edge roll Dressing does not adhere to the wound bed and peri-wound skin (undisturbed healing environment) Can be worn for up to 7 days on skin and compromised skin 1½ x 1½ inch pad