Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 3-1/2 X 3-1/2 inch With Border Film Backing Acrylic Adhesive Square Sterile, 90610, Each
Catalog No. 502831249
Encompass_Preferred
3M's Tegaderm™ high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering The white foam dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This 3½ by 3½-inch square dressing will not stick to wounds or disrupt healing tissue
Catalog No. 50-283-1249 Supplier Med Vet International Supplier No. 465140EA
3M's Tegaderm™ high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering The white foam dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This 3½ by 3½-inch square dressing will not stick to wounds or disrupt healing tissue