Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 5-5/8 X 5-5/8 inch With Border Film Backing Acrylic Adhesive Square Sterile, 90612, 10/Box
Catalog No. 502830796
Encompass_Preferred
3M's Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system and square shape The thin film backing blocks bacteria and viruses from entering The Tegaderm dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This dressing will not stick to wounds or disrupt healing tissue 14.3 x 14.3 centimeters
Catalog No. 50-283-0796 Supplier Med Vet International Supplier No. 459259BX
3M's Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system and square shape The thin film backing blocks bacteria and viruses from entering The Tegaderm dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This dressing will not stick to wounds or disrupt healing tissue 14.3 x 14.3 centimeters