Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 4 X 4 inch Without Border Film Backing NonAdhesive Square Sterile, 90601, 10/Box
Catalog No. 502829305
Encompass_Preferred
Provides a water resistant seal while dressing is intact Allows showering Dressing absorbs/manages low to high levels of exudate, protects peri-wound skin and maintains moisture balance Gentle, absorbent and secure silicone foam border dressing easily handles high to low exudating wounds Stays in place, yet removes gently, making it a good choice for fragile skin Adhesive film border is thin and has a low profile to minimize edge roll Dressing does not adhere to the wound bed and peri-wound skin (undisturbed healing environment) Can be worn for up to 7 days on skin and compromised skin 4 x 4 inch dressing with 2-3/8 x 2-3/8 inch pad
Catalog No. 50-282-9305 Supplier Med Vet International Supplier No. 688562BX
Provides a water resistant seal while dressing is intact Allows showering Dressing absorbs/manages low to high levels of exudate, protects peri-wound skin and maintains moisture balance Gentle, absorbent and secure silicone foam border dressing easily handles high to low exudating wounds Stays in place, yet removes gently, making it a good choice for fragile skin Adhesive film border is thin and has a low profile to minimize edge roll Dressing does not adhere to the wound bed and peri-wound skin (undisturbed healing environment) Can be worn for up to 7 days on skin and compromised skin 4 x 4 inch dressing with 2-3/8 x 2-3/8 inch pad