Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm Hp 4 X 4-3/4 inch Frame Style DelIVery Rectangle Sterile, 9536Hp, 50/Box
Catalog No. 502830303
Encompass_Preferred
High adhesion keeps dressings in place under challenging conditions with up to 7-day wear time Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Transparency allows for easy monitoring of the wound or I.V. site without the need to remove the dressing Picture-frame delivery makes placement accurate and easy, minimizing potential to stick to gloves or to itself Dressing is conformable and flexes with skin for greater patient comfort Oval dressing is 4 x 4¾ inches
Catalog No. 50-283-0303 Supplier Med Vet International Supplier No. 210037BX
High adhesion keeps dressings in place under challenging conditions with up to 7-day wear time Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Transparency allows for easy monitoring of the wound or I.V. site without the need to remove the dressing Picture-frame delivery makes placement accurate and easy, minimizing potential to stick to gloves or to itself Dressing is conformable and flexes with skin for greater patient comfort Oval dressing is 4 x 4¾ inches