Learn More
Med Vet International IV Dressing 3M Tegaderm Film 4 X 4-3/4 inch Sterile, 1688, 50/Box
Catalog No. 502831004
Encompass_Preferred
Cost-effective I.V. and central line catheter securement solution Designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture Innovative adhesives balance securement and gentle removal Easy to apply and remove Safe to wear up to 7 days for CVCs and may be worn for the life of the PIV catheter Waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses 4 x 4¾ inches
Catalog No. 50-283-1004 Supplier Med Vet International Supplier No. 764489BX
Cost-effective I.V. and central line catheter securement solution Designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture Innovative adhesives balance securement and gentle removal Easy to apply and remove Safe to wear up to 7 days for CVCs and may be worn for the life of the PIV catheter Waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses 4 x 4¾ inches