Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm Hp 2-3/8 X 2-3/4 inch Frame Style DelIVery Rectangle Sterile, 9534Hp, 100/Box
Catalog No. 502830301
Encompass_Preferred
High adhesion keeps dressings in place under challenging conditions with up to 7-day wear time Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Transparency allows for easy monitoring of the wound or I.V. site without the need to remove the dressing Picture-frame delivery makes placement accurate and easy, minimizing potential to stick to gloves or to itself Dressing is conformable and flexes with skin for greater patient comfort Octagonal dressing is 2-3/8 x 2-3/4 inches
Catalog No. 50-283-0301 Supplier Med Vet International Supplier No. 210036BX
High adhesion keeps dressings in place under challenging conditions with up to 7-day wear time Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Transparency allows for easy monitoring of the wound or I.V. site without the need to remove the dressing Picture-frame delivery makes placement accurate and easy, minimizing potential to stick to gloves or to itself Dressing is conformable and flexes with skin for greater patient comfort Octagonal dressing is 2-3/8 x 2-3/4 inches