Learn More
Med Vet International Alginate Dressing 3M Tegaderm High integrity 12 inch Length Rope, 90120, 5/Box
Catalog No. 502831366
Encompass_Preferred
Conformable: Gel forming dressing primary dressing Supports healing: Maintains optimal moisture levels needed for healing tissue Absorbent: For use on medium to high exuding wounds Compatible with 3M cover dressings
Catalog No. 50-283-1366 Supplier Med Vet International Supplier No. 318723BX
Conformable: Gel forming dressing primary dressing Supports healing: Maintains optimal moisture levels needed for healing tissue Absorbent: For use on medium to high exuding wounds Compatible with 3M cover dressings