Learn More
Med Vet International Foam Dressing 3M Tegaderm 6 X 6 inch With Border Film Backing Silicone Adhesive Square Sterile, 90642, 40/Case
Catalog No. 502831686
Encompass_Preferred
Provides a water resistant seal while dressing is intact Allows showering Dressing absorbs/manages low to high levels of exudate, protects peri-wound skin and maintains moisture balance Gentle, absorbent and secure silicone foam border dressing easily handles high to low exudating wounds Stays in place, yet removes gently, making it a good choice for fragile skin Adhesive film border is thin and has a low profile to minimize edge roll Dressing does not adhere to the wound bed and peri-wound skin (undisturbed healing environment) Can be worn for up to 7 days on skin, including compromised skin 4-1/8 x 4-1/8 inch pad
Catalog No. 50-283-1686 Supplier Med Vet International Supplier No. 842346CS
Provides a water resistant seal while dressing is intact Allows showering Dressing absorbs/manages low to high levels of exudate, protects peri-wound skin and maintains moisture balance Gentle, absorbent and secure silicone foam border dressing easily handles high to low exudating wounds Stays in place, yet removes gently, making it a good choice for fragile skin Adhesive film border is thin and has a low profile to minimize edge roll Dressing does not adhere to the wound bed and peri-wound skin (undisturbed healing environment) Can be worn for up to 7 days on skin, including compromised skin 4-1/8 x 4-1/8 inch pad