Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 7-7/8 X 8 inch Frame Style DelIVery Square Sterile, 90805, 40/Case
Supplier:  Med Vet International 1210644CS
Encompass_Preferred
Rectangular transparent dressing is indicated for partial and full thickness dermal ulcers; skin tears; pressure ulcers; superficial wounds and abrasions; superficial partial-thickness burns; donor sites; and clean, closed approximated surgical incisions or laparoscopic incisions Transparent film allows wound or surgical incision to be monitored without dressing removal Clear design takes the guesswork out of application over the wound or surgical incision Novel acrylic polymer pad is breathable and designed to absorb low to moderate exudate to promote an optimal healing environment Dressing does not break down in the wound or surgical incision Nonbulky design helps to prevent dressing roll or lift due to friction from linens, clothing or prosthetics Protects from external contaminants, bodily fluids, bacteria, viruses and water ingress Wear time of up to 7 days gives for extended protection beyond the initial hospital stay and into the home
Catalog No. 50-283-2346