Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 2-3/4 X 2-3/4 inch With Border Film Backing Acrylic Adhesive Oval Sterile, 90614, Each
Catalog No. 502830606
Encompass_Preferred
3M's Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering The oval foam dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This 6.9 x 7.6 centimeter dressing will not stick to wounds or disrupt healing tissue
Catalog No. 50-283-0606 Supplier Med Vet International Supplier No. 695850EA
3M's Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering The oval foam dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This 6.9 x 7.6 centimeter dressing will not stick to wounds or disrupt healing tissue