Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 5-1/2 X 5-1/2 inch With Border Film Backing Acrylic Adhesive Heel Sterile, 90619, Each
Catalog No. 502832320
Encompass_Preferred
Multi-layer design provides high absorbency with high breathability to reduce the risk of maceration Adapts to changing levels of exudate to maintain moisture balance for optimal wound healing Particularly suited for situations where increased securement or longer wear time is needed Conformable for difficult body contours 3 x 3 inch pad
Catalog No. 50-283-2320 Supplier Med Vet International Supplier No. 472010EA
Multi-layer design provides high absorbency with high breathability to reduce the risk of maceration Adapts to changing levels of exudate to maintain moisture balance for optimal wound healing Particularly suited for situations where increased securement or longer wear time is needed Conformable for difficult body contours 3 x 3 inch pad