Learn More
Med Vet International Foam Dressing 3M Tegaderm 7-1/4 X 8-3/4 inch With Border Film Backing Silicone Adhesive Large Sacral Sterile, 90648, Each
Catalog No. 502830233
Encompass_Preferred
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 7-1/4 x 8-3/4 inch sacral shape is good for lower back applications
Catalog No. 50-283-0233 Supplier Med Vet International Supplier No. 1078889EA
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 7-1/4 x 8-3/4 inch sacral shape is good for lower back applications