Learn More
Med Vet International IV Dressing 3M Tegaderm Film 2-1/2 X 2-3/4 inch Sterile, 1683, 400/Case
Catalog No. 502831590
Encompass_Preferred
Designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture Cost-effective I.V. and central line catheter securement solution Innovative adhesives balance securement and gentle removal Easy to apply and remove Safe to wear up to 7 days for CVCs and may be worn for the life of the PIV catheter Waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses
Catalog No. 50-283-1590 Supplier Med Vet International Supplier No. 761469CS
Designed to maintain securement by preventing edge lift, flexing with patient movement and managing moisture Cost-effective I.V. and central line catheter securement solution Innovative adhesives balance securement and gentle removal Easy to apply and remove Safe to wear up to 7 days for CVCs and may be worn for the life of the PIV catheter Waterproof film coating on all pieces provides a barrier to external contaminants including liquids, bacteria and viruses